Learn More
Medchemexpress LLC 2,4-pyridine dicarboxylic acid | 499-80-9 | MFCD00006296 | 99.8% | 167.12 g/mol | C7H5NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01713210MM/1ML
Encompass_Preferred
2,4-pyridine dicarboxylic acid (2,4-PDCA) is a small-molecule inhibitor of 2-oxoglutarate (2OG)-dependent oxygenases, including JmjC histone demethylases. It is offered for laboratory research as a high-purity solid and as a ready-to-use 10 mM solution in DMSO, and is commonly used in epigenetics and enzymology studies.
  • Broad-spectrum 2OG oxygenase inhibitor suitable for demethylase assays.
  • High purity suitable for biochemical and cellular studies.
  • Available as a ready-to-use 10 mM solution in DMSO for convenience.
  • Solid form available for custom formulation and scale-up.
  • Documented identifiers and safety data for research compliance.
Catalog No. 50-003-00341