Learn More
Medchemexpress LLC 2,4-Pyrimidinediamine, 5-[(3,4-dimethoxyphenyl)methyl]- | 5355-16-8 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1902500MG
Encompass_Preferred
Diaveridine is an antibacterial agent and a dihydrofolate reductase (DHFR) inhibitor, demonstrating a Ki of 11.5 nM for the wild-type DHFR. For research use only, this product is not intended for patient use. In vitro treatments with Diaveridine for 90 minutes show a strong bactericidal effect on S. typhimurium TA1535, with no bacterial growth observed at concentrations of 10 μg/mL or more.
  • Dihydrofolate reductase (DHFR) inhibitor
  • Antibacterial agent
  • Ki of 11.5 nM for wild-type DHFR
  • Strong bactericidal effect on S. typhimurium TA1535 at ≥ 10 μg/mL
Catalog No. 50-004-11265