Learn More
Medchemexpress LLC 2,4-Pyrimidinediamine, N2-[4-[(3-aminopropyl)amino]phenyl]-N4-(5-cyclopropyl-1H-pyrazol-3-yl) HCl | 2700435-52-3 | 99.5% | 400.91 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY18625A50MG
Encompass_Preferred
VEGFR-2-IN-5 hydrochloride is an off-white to light yellow solid. It is identified as a VEGFR2 inhibitor, extracted from patent WO2013055780A1. This compound targets VEGFR, demonstrating activity against VEGFR2.
  • Acts as a VEGFR2 inhibitor.
  • Appearance: off-white to light yellow solid.
  • Complies with given specifications.
Catalog No. 50-003-87884