Learn More
Medchemexpress LLC 2,4-Thiazolidinedicarboxylic acid | 30097-06-4 | MFCD00145399 | 99.8% | 177.18 | C5H7NO4S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0242415G
Encompass_Preferred
Tidiacic (thiazolidine-2,4-dicarboxylic acid) is a hepatoprotective research compound that acts as an antioxidant and sulfur donor. It is supplied as a solid with analytical characterization suitable for laboratory studies of liver protection, oxidative stress, and related biochemical assays. For research use only.
  • Acts as an antioxidant and sulfur donor in biological studies.
  • Suitable for in vitro and biochemical assays.
  • High purity (≈99.8%).
  • Solid form, off-white to light-yellow, facilitating handling and storage.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-003-01617