Learn More
Medchemexpress LLC 2-(4-trifluoromethyl-phenylamino)benzoic acid | 1566-81-0 | 99.9% | 324.25 g·mol⁻1 | C15H11F3N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10024425MG
Encompass_Preferred
NS1652 is a reversible anion conductance inhibitor that blocks chloride channels. It potently inhibits chloride conductance in human and mouse red blood cells (IC50 = 1.6 μM) and shows weak activity against volume-regulated anion channels (IC50 ≈ 125 μM). Supplied as a high-purity research reagent for biochemical and cellular studies.
  • Reversible anion conductance inhibitor.
  • Potent chloride channel inhibition (IC50 1.6 μM in human and mouse red blood cells).
  • Weak VRAC inhibition (IC50 ≈ 125 μM) enabling selectivity studies.
  • High purity suitable for research applications.
  • Available in small quantities for experimental use.
  • Useful for studying anion conductance and erythrocyte physiology.
Catalog No. 50-002-15625