Learn More
Novus Biologicals 2-40 kDa Peggy Sue or Sally Sue Separation Module
SDP
Supplier:  Novus Biologicals SMS003
Encompass
Requires Compass v3.0 or higher. Use with a Detection Module. Reagents for 480 data points includes two parts: Separation box with 5 boxes of Capillaries (96 capillaries per box), 5 Plates, Wash Buffer, Upper and Lower Running Buffers 4, Upper and Lower Running Buffer cups, Wash Buffer cup, 5 Manifold Cleaning Sponges, Separation Matrix 2, Stacking Matrix 2, 10X Sample Buffer, plus 2 x Standard Pack 5 clamshell with 8 ea. DTT, FL Standard master mix with 26 kDa System Control protein (Store at 4°C).
Catalog No. NC1177845