Learn More
Medchemexpress LLC 2,5(1H,6H)-quinolinedione, 7,8-dihydro | 15450-69-8 | MFCD00766821 | 99.9% | 163.17 g/mol | C9H9NO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01776410G
Encompass_Preferred
7,8-Dihydro-2,5(1H,6H)-quinolinedione (compound SF32) is a research-grade small molecule described in literature as a positive inotropic agent. It is supplied as a solid with high analytical purity and intended for laboratory research applications; refer to the manufacturer documentation and certificate of analysis for handling, storage, and stability details.
  • High reported purity: 99.86%.
  • Solid form; appearance light brown to light yellow.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Solvent storage guidance: -80°C for 6 months, -20°C for 1 month.
  • Suitable for in vitro research as a positive inotropic agent.
  • Available in multiple pack sizes for laboratory use.
  • Supplied with a certificate of analysis for quality verification.
Catalog No. 50-002-99719