Learn More
Medchemexpress LLC 2,5-anhydro-D-mannitol | 41107-82-8 | MFCD00012443 | 97.0% | 164.16 g·mol⁻1 | C6H12O5 | 5 MG

Supplier: Medchemexpress LLC HYW0393155MG
2,5-Anhydro-D-mannitol (CAS 41107-82-8) is a small-molecule biochemical assay reagent used in metabolic and enzymatic studies. It is supplied as a solid with high stated purity and is suitable as a reference standard or reagent in biochemical assays.
- Purity 97%.
- Molecular weight 164.16 g·mol⁻1.
- Chemical formula C6H12O5.
- Appearance solid.
- Typical applications biochemical assays and enzymatic studies.
- Available in small milligram quantities for analytical and assay use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.