Learn More
Medchemexpress LLC 2-((5-bromothiophen-2-yl)methyleneamino)benzamide | 304456-62-0 | MFCD00977636 | 98.0% | 309.18 g/mol | C12H9BrN2OS | 50MG
SDP
Supplier:  Medchemexpress LLC HY12444750MG
Encompass_Preferred
BTYNB is a small-molecule inhibitor of IMP1 that disrupts IMP1 binding to c-Myc mRNA and is used in biochemical and cellular research. The compound is supplied as a white to light yellow solid with reported high purity and characterized molecular properties.
  • Potent IMP1 inhibitor that disrupts IMP1-c-Myc mRNA binding (reported IC50 = 5 μM).
  • High purity solid suitable for research applications (reported purity 98.0%).
  • Well-characterized molecular properties (formula C12H9BrN2OS, MW 309.18 g/mol).
  • Stable when stored protected from light and refrigerated as recommended.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-37081