Learn More
Medchemexpress LLC 2,5-cyclohexadien-1-one, 2,6-dichloro-4-[(4-hydroxyphenyl)imino]-, sodium salt (1:1) | 620-45-1 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0018100MG
Encompass_Preferred
DCIP sodium is a blue dye commonly used in various biochemical and biotechnological applications as an indicator of redox reactions. It possesses unique chemical properties that enable it to change color based on the oxidation state of the substance being tested. This reagent is frequently employed in enzyme assays, such as for measuring the activity of succinate dehydrogenase, and in protein quantification methods, including the Lowry assay. DCIP sodium also serves as a biochemical reagent for life science-related research.
  • Blue dye
  • Acts as an indicator of redox reactions
  • Changes color based on the oxidation state of the tested substance
  • Utilized in enzyme assays (e.g., succinate dehydrogenase activity measurement)
  • Applied in protein quantification methods (e.g., Lowry assay)
  • Suitable as a biochemical reagent for life science research
Catalog No. 50-004-10808