Learn More
Medchemexpress LLC 2,5-Cyclohexadiene-1,4-dione, 2-[(1,4-dimethylpentyl)amino]-5-(phenylamino)- | 2894124-00-4 | 98.9% | 312.41 g/mol | C19H24N2O2 | 10 MG

Supplier: Medchemexpress LLC HY16788710MG
7PPD-Q is a substituted p-phenylenediamine antioxidant quinone derivative used in environmental toxicology and analytical studies; it has demonstrated toxicity to the bacterium V. fischeri (EC50 = 14.9 mg/L).
- Provides a quinone derivative of 7-PPD for environmental and metabolite studies.
- High purity (98.88%).
- Molecular formula C19H24N2O2; molecular weight 312.41 g/mol.
- Solubility in DMSO up to 50 mg/mL; in vivo formulation example 10% DMSO/90% corn oil ≥2.5 mg/mL.
- Appearance: pale purple to purple solid.
- Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
- Supplied in small mass quantities suitable for laboratory assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.