Learn More
Medchemexpress LLC 2,5-Cyclohexadiene-1,4-dione, 2,5-bis[(2-methylphenyl)amino]- | 252950-56-4 | 99.7% | 318.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16365050MG
Encompass_Preferred
DTPD-Q is a transformation product of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and increases ecological risk in the environment. This product is intended for research use only and is not sold to patients.
  • Transformation product of a common tire additive.
  • Accumulates and raises ecological risk in the environment.
  • High purity of 99.68%.
Catalog No. 50-003-91942