Learn More
Medchemexpress LLC 2,5-cyclohexadiene-1,4-dione, 2,5-bis(phenylamino)- | 3421-08-7 | >98.0% | 290.32 g/mol | C18H14N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW1761995MG
Encompass_Preferred
DPPD-Q is an oxidized derivative of the antioxidant substituted p-phenylenediamine DPPD. Supplied as a solid research reagent with high purity, it is used as a jack bean urease inhibitor and in studies of reactive oxygen species and environmental contaminants.
Catalog No. 50-002-81586