Learn More
Medchemexpress LLC 2,5-Dibromohexanediamide | 99584-96-0 | MFCD27976335 | >95.0% | 301.96 g/mol | C6H10Br2N2O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1018475G
Encompass_Preferred
DBHDA (2,5-dibromohexanediamide) is an organic reagent used to convert cysteine residues into dehydroalanine (Dha) in peptides and proteins. It is applied in synthetic biology and peptide/protein modification workflows to enable site-selective transformations and downstream conjugation chemistries.
  • Converts cysteine residues to dehydroalanine for site-selective modification.
  • Enables selective, mild conversion compatible with peptide chemistry.
  • Supplied as an off-white solid for bench-top use.
  • Typical purity reported ≥95%, suitable for research applications.
  • Facilitates downstream conjugation and functionalization of peptides and proteins.
Catalog No. 50-002-48138