Learn More
Medchemexpress LLC 2',5'-dideoxyadenosine | 6698-26-6 | MFCD00210904 | 99.9% | C10H13N5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13587810MG
Encompass_Preferred
2',5'-Dideoxyadenosine is a nucleoside analog and a potent, non-competitive inhibitor of adenylyl cyclase that binds the P-site. It is used as a research reagent to probe cAMP signaling, adenylyl cyclase activity, and downstream phosphorylation events in vitro and in vivo. Supplied as a solid with high purity and recommended storage for stability.
  • Potent, non-competitive adenylyl cyclase inhibitor (IC50 = 3 μM).
  • Reduces cAMP production and modulates phosphorylation of signaling proteins such as GluA1 and Akt.
  • Effective in vitro and in vivo for studying antiadrenergic and diuretic/natriuretic responses.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when stored as powder at recommended temperatures for extended periods.
Catalog No. 50-002-61482