Learn More
Medchemexpress LLC 2,5-Dihydroxy-1,4-benzoquinone | 615-94-1 | 99.95% | 140.09 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0103231G
Encompass_Preferred
2,5-Dihydroxy-1,4-benzoquinone (DHBQ) is a redox-active quinone-based organic ligand with antibacterial activity, obtainable from fungal metabolites. It can be used to construct conductive metal-organic frameworks for Li+-ion batteries and enhances Li+-ion battery cathode discharge capacity when incorporated into the Fe (dhbq) metal-organic framework.
  • Redox-active quinone-based organic ligand
  • Exhibits antibacterial activity
  • Originates from fungal metabolites
  • Enhances Li+-ion battery cathode discharge capacity
  • Used to construct conductive metal-organic frameworks for Li+-ion batteries
  • Functions as a redox-active cathode material for lithium-ion batteries (LIBs)
Catalog No. 50-004-53892