Learn More
Medchemexpress LLC 2,5-dioxo-1-pyrrolidinyl 11-azidoundecanoate | 850080-13-6 | MFCD29041759 | 98.0% | 324.38 g/mol | C15H24N4O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1909261G
Encompass_Preferred
2,5-Dioxo-1-pyrrolidinyl 11-azidoundecanoate is an azide-containing N-hydroxysuccinimide (NHS) ester linker for bioconjugation. It provides an activated carboxylate for efficient coupling to primary amines and a terminal azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition. Typical applications include linker synthesis, protein and peptide labeling, and assembly of multifunctional conjugates.
  • Activated NHS ester for efficient amine coupling
  • Terminal azide for click-chemistry conjugation
  • High purity (98.0% by NMR)
  • Chemical formula C15H24N4O4, MW 324.38 g/mol
  • Available as a solid; common package 1 G
Catalog No. 50-002-81622