Learn More
Medchemexpress LLC 2,5-dioxo-1-pyrrolidinyl 11-azidoundecanoate | 850080-13-6 | MFCD29041759 | 98.0% | 324.38 g/mol | C15H24N4O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW190926500MG
Encompass_Preferred
2,5-Dioxo-1-pyrrolidinyl 11-azidoundecanoate is an azide-containing N-hydroxysuccinimide (NHS) ester linker for bioconjugation. The molecule couples to primary amines through the NHS ester and enables attachment via azide-alkyne click chemistry, making it suitable for labeling, probe synthesis, and linker assembly in chemical biology applications.
  • Contains an azide group for click chemistry (CuAAC and strain-promoted reactions).
  • Contains an NHS ester reactive toward primary amines for stable conjugation.
  • High purity for reliable conjugation and analytical reproducibility.
  • Available in laboratory-scale quantities for synthesis and labeling workflows.
  • Handle under anhydrous conditions and store cold to preserve activity.
Catalog No. 50-002-81943