Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 1-(4-(1,2,4,5-tetrazin-3-yl)phenyl)-3,19-dioxo-6,9,12,15-tetraoxa-22,23-... | 95.0% | 723.82 | C30H41N7O10S2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1360405MG

Encompass_Preferred

Tetrazine-PEG4-SS-NHS is a cleavable PEG4 linker that features a tetrazine reactive group, a disulfide (SS) cleavable spacer, and an N-hydroxysuccinimide (NHS) ester for efficient bioconjugation and click chemistry applications.

  • Cleavable disulfide spacer enables reductive release of payloads.
  • Tetrazine group allows inverse electron demand Diels-Alder (iEDDA) click reactions.
  • N-hydroxysuccinimide (NHS) ester provides amine-targeted conjugation chemistry.
  • PEG4 spacer improves solubility and provides flexibility between conjugated partners.
  • Typical purity 95.0% by NMR.
  • Molecular weight 723.82, solid, purple to purplish red in color.
  • Storage: powder at -20 °C; in solvent store at -80 °C (up to 6 months) or -20 °C (up to 1 month).

Catalog No. 50-002-03822


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.