Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23,26,29,32,35,38-tridecaoxahentetracontan-41-oate | 3063847-98-0 | 95.0% | C32H59NO17 | 10MG

Supplier: Medchemexpress LLC HY14110710MG
m-PEG13-NHS ester is a PEG-based reactive linker containing an N-hydroxysuccinimide (NHS) ester for amine-selective coupling. It is intended for use as a spacer in the synthesis of PROTACs and other bioconjugation applications where a flexible, hydrophilic linker is required.
- PEG13 spacer providing flexibility and hydrophilicity.
- NHS ester for amine-selective coupling.
- High purity (95.0%).
- Soluble in DMSO at 100 mg/mL; ultrasonic treatment may be required.
- Oil, colorless to off-white appearance.
- Store sealed at -20°C; in solution store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.