Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulfanyl)butanoate | 107348-47-0 | MFCD28398150 | 98.0% | 326.40 g/mol | C13H14N2O4S2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW07100650MG

Encompass_Preferred

PPC-NHS ester is a cleavable linker reagent containing an N-hydroxysuccinimide (NHS) ester and a pyridin-2-yldisulfanyl cleavable moiety; it is used to attach cytotoxic payloads to antibodies and enable payload release under reducing conditions.

  • Cleavable disulfide linker enabling intracellular release of payloads
  • N-hydroxysuccinimide (NHS) ester reactive toward primary amines
  • Molecular formula C13H14N2O4S2, molecular weight 326.40 g/mol
  • Purity 98.0%
  • Supplied as a white to off-white solid, 50 MG per package
  • Storage: powder -20°C (stable 3 years); in solvent -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-03095


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.