Learn More
Medchemexpress LLC 2,5-Piperazinedione, 3-(1H-indol-3-ylmethyl)-6-(2-methylpropyl)-, (3S,6S)- | 15136-34-2 | 99.90% | 299.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP2092
Encompass_Preferred
Cyclo(L-leucyl-L-tryptophyl), also known as Cyclo(Leu-Trp), is a cyclic dipeptide that acts as a melatonin receptor agonist and a bitter ligand. This compound has been observed to inhibit a variety of bacteria and fungi, showcasing its potential in anti-infection pathways. Its formal chemical name is 2,5-Piperazinedione, 3-(1H-indol-3-ylmethyl)-6-(2-methylpropyl)-, (3S,6S)-.
  • Inhibits various bacteria and fungi.
  • Acts as a melatonin receptor agonist.
  • Used as a bitter ligand.
  • Involved in anti-infection, GPCR/G Protein, and neuronal signaling pathways.
  • Recommended storage conditions for powder are sealed storage, away from moisture, at -80°C for up to 2 years or -20°C for 1 year.
  • For solutions in solvent, recommended storage is sealed storage, away from moisture, at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-26788