Learn More
Medchemexpress LLC 2,5-Piperazinedione, 3-[(4-hydroxyphenyl)methyl]-6-(2-methylpropyl)- | 82863-65-8 | MFCD21333410 | 98.9% | 276.34 g/mol | C15H20N2O3 | 5 MG

Supplier: Medchemexpress LLC HY1311155MG
Cyclo(Tyr-Leu) is a cyclic dipeptide composed of tyrosine and leucine, commonly used as a research reagent in biochemical and pharmacological studies. It is supplied as a purified analytical sample with supporting documentation for experimental use.
- High purity (98.9%).
- Molecular weight 276.34 g/mol.
- Chemical formula C15H20N2O3.
- Available in small quantities suitable for assay work, e.g. 5 mg vials.
- Analytical data provided: COA, LCMS, HNMR, SDS.
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.