Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterprise Technology Solutions 2.5 W H2O Fire Extinguisher
SDP

Supplier:  Enterprise Technology Solutions KID466403

Encompass_Preferred

ProPlus 2 5 W H2O Fire Extinguisher 2-A 2 5 gal 20 86 lb

Catalog No. 50-302-5623


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.