Learn More
Business Communication Solutions 2.50"W x 3.50"L, Blank White Polyester Label, Permanent Adhesive, Blank, Horizontal Perforations Between Labels, 1730 Labels/Roll, Wound out on a 3.00" Core, 8.00" OD Roll.
Supplier:  Business Communication Solutions 10009843Z
Encompass_Preferred
2.50"W x 3.50"L, Blank White Polyester Label, Permanent Adhesive, Blank, Horizontal Perforations Between Labels, 1730 Labels/Roll, 3.00" Core, 8.00" OD Roll. Compatible with the Zebra industrial printers. Compatible with all full size industrial thermal printers including Zebra, Honeywell, Datamax and Sato. One year expiration.
Catalog No. NC2200733