Learn More
Electronic Imaging Materials Inc 2.52 Inches x 74 Meter T68-G Premium Resin Ribbon - EIMINC
SDP
Supplier:  Electronic Imaging Materials Inc T68252074G
Encompass_Preferred
T68-G—2.52 inches x 74 meter resin ribbon specially formulated to print exceptional images. Handles some of the harshest environmental conditions—including resistance against abrasion, heat, steam and chemicals such as gasoline, mineral spirits, oil, bleach, IPA, antifreeze, and sulfuric acid. *Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra GX430 series printers, as well as TSC TC 200/210/300/310 printers.
Catalog No. NC1192024