Learn More
US Plastic Corporation 2.5L SQUARE CARBOY HDPE
SDP
Supplier:  US Plastic Corporation 74093
Encompass
This high density polyethylene carboy meets FDA standards. Features a screw closure with a center 3/4 in. NPT reducer in cap and also, a carrying handle. Can easily see the liquid level due to this carboy being translucent.
Catalog No. NC9945358