Learn More
Medchemexpress LLC 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, hydrochloride (1:2), (6S)- | 104632-25-9 | 98.0% | 284.25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17355100MG
Encompass_Preferred
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. It can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist.
  • Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively.
  • Applicable for research into Parkinson's disease (PD) and restless legs syndrome (RLS).
Catalog No. 50-003-84989