Learn More
Medchemexpress LLC 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, hydrochloride (1:2), (6S)- | 104632-25-9 | 99.7% | 284.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1735550MG
Encompass_Preferred
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It has Ki values of 2.2 nM for the D2-type receptor, 3.9 nM for D2, 0.5 nM for D3, and 1.3 nM for D4 receptors. It is used in the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist
  • Can be used for research of Parkinson's disease (PD)
  • Can be used for research of restless legs syndrome (RLS)
Catalog No. 50-003-84620