Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-Dichloro-4-pyridinamine | 2587-02-2 | 163.007 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0081991G

Encompass_Preferred

2,6-Dichloro-4-pyridinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • High purity of 100.0%
  • Available in solid form
  • White to light yellow appearance
  • Suitable for life science research
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-46188


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.