Learn More
Medchemexpress LLC 2,6-Dichloro-4-pyridinamine | 2587-02-2 | 163.007 | 1 ML

Supplier: Medchemexpress LLC HYW00819910MM/1ML
2,6-Dichloro-4-pyridinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
- Purity: 99.98%
- Molecular weight: 163.007
- Appearance: White to light yellow solid
- CAS number: 2587-02-2
- Molecular formula: C5H4Cl2N2
- Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.