Learn More
Medchemexpress LLC 2,6-dichloro-N-(2,6-dimethylphenyl)hexanamide | 1037184-07-8 | MFCD25562938 | >98.0% | 288.21 g/mol | C14H19Cl2NO | 5 MG

Supplier: Medchemexpress LLC HYW3285205MG
2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide is an amide impurity associated with the local anesthetic bupivacaine. Supplied for research and analytical use in small milligram quantities, the substance is used as an impurity reference standard and for method development and quality control in laboratories.
- CAS number: 1037184-07-8
- Chemical formula: C14H19Cl2NO
- Molecular weight: 288.21 g/mol
- Purity: >98.0%
- Typical pack size: 5 mg for research applications
- Common use: impurity reference for analytical and method development
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.