Learn More
Medchemexpress LLC 2,6-dichloro-N-[2-(cyclopropanecarbonylamino)-4-pyridinyl]benzamide | 1258292-64-6 | >98.0% | 25 MG

Supplier: Medchemexpress LLC HY12046925MG
2,6-dichloro-N-(2-(cyclopropanecarbonylamino)-4-pyridinyl)benzamide is a small-molecule, orally bioavailable TYK2 inhibitor used in preclinical research to probe TYK2 and JAK family kinase biology.
- Potent TYK2 inhibition with reported Ki = 4.8 nM.
- Reported JAK family kinase activity: JAK1 = 0.7 nM, JAK2 = 0.7 nM, JAK3 = 0.4 nM.
- Molecular formula C16H13Cl2N3O2, molecular weight 350.2 g/mol.
- High purity ≥98% by HPLC.
- Appearance: solid, off-white to yellow; supplied as a powder for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.