Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-Dimethoxybenzoic acid | 1466-76-8 | 99.9% | C9H10O4 | 10 MM 1 ML
SDP

Supplier:  Medchemexpress LLC HY7650410MM/1ML

Encompass_Preferred

2,6-Dimethoxybenzoic acid is an organic compound classified as an o-methoxybenzoic acid derivative. It acts as an endogenous metabolite and is commonly used in research studies. This product is suitable for various laboratory applications and requires specific storage conditions to maintain stability.

  • CAS number: 1466-76-8
  • Molecular formula: C9H10O4
  • Molecular weight: 182.17
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease

Catalog No. 50-004-34099


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.