Learn More
Medchemexpress LLC 2-(6-methoxynaphthalen-2-yl)acetic acid | 23981-47-7 | MFCD00033105 | 216.23 g/mol | C13H12O3 | 10 MG

Supplier: Medchemexpress LLC HYW08689610MG
α-Demethylnaproxen (2-(6-methoxynaphthalen-2-yl)acetic acid) is an analytical reference standard and the major metabolite of nabumetone, supplied for research and analytical applications.
- Analytical standard for metabolite identification and impurity testing.
- Major metabolite of nabumetone; used as a reference in metabolic studies.
- CAS number 23981-47-7 and MFCD00033105 for unambiguous identification.
- Molecular formula C13H12O3 and molecular weight 216.23 g/mol.
- Supplied in small analytical quantities appropriate for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.