Learn More
Medchemexpress LLC 2-[6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic acid | 189005-44-5 | MFCD04114705 | 99.8% | 280.32 | C17H16N2O2 | 50 G

Supplier: Medchemexpress LLC HY10822850G
Zolpidic acid is an imidazo[1,2-a]pyridine-3-acetic acid derivative (CAS 189005-44-5) used as a chemical intermediate and precursor in the synthesis of zolpidem. Supplied as a high-purity solid for research and synthetic chemistry, it provides a stable imidazopyridine acetic acid building block for laboratory-scale synthesis and analytical work.
- High purity solid suitable for synthetic use.
- Molecular weight 280.32 and formula C17H16N2O2.
- Commonly used as a precursor in zolpidem synthesis.
- Available in a range of lab-scale pack sizes.
- Soluble in common organic solvents such as DMF and DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.