Learn More
Medchemexpress LLC 2,6-Piperidinedione, 3-(4-bromo-1,3-dihydro-1-oxo-2H-isoindol-2-yl)- | 2093387-36-9 | 99.86% | 323.14 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY43722
Encompass_Preferred
Lenalidomide-Br (Compound 41) is an analog of cereblon (CRBN) ligand Lenalidomide for E3 ubiquitin ligase, utilized in the recruitment of CRBN protein. It can be linked to a ligand for protein by a linker to create PROTACs.
  • Analog of cereblon (CRBN) ligand for E3 ubiquitin ligase
  • Used in the recruitment of CRBN protein
  • Can be connected to form PROTACs, which are inducers of ubiquitination-mediated degradation of cancer-promoting proteins
Catalog No. 50-004-02891