Learn More
Medchemexpress LLC 2,6-piperidinedione, 3-(4-bromo-2-fluorophenyl) | 2924092-66-8 | 98.6% | 286.10 | C11H9BrFNO2 | 100mg
SDP
Supplier:  Medchemexpress LLC HYW888970100MG
Encompass_Preferred
3-(4-Bromo-2-fluorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC
Catalog No. 50-002-68320