Learn More
Medchemexpress LLC 2,6-piperidinedione, 3-(4-bromo-7-fluoro-1,3-dihydro-1-oxo-2H-isoindol-2-yl)- | 2438239-34-8 | MFCD34576717 | 97.0% | 341.13 g/mol | C13H10BrFN2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW46022725MG
Encompass_Preferred
Lenalidomide-4-Br-7-fluoro is a synthetic derivative of lenalidomide bearing bromine and fluorine substituents on the benzyl ring. It is intended for research use and is employed as a ligand for E3 ubiquitin ligases and in PROTAC-related studies. Molecular formula C13H10BrFN2O3; molecular weight 341.13 g/mol.
  • Derivative of lenalidomide with bromine and fluorine substituents.
  • Used as an E3 ligase ligand in PROTAC design and target degradation research.
  • Certificate of analysis reports purity of 97.0% (NMR).
  • Supplied as a powder with recommended storage: -20°C (3 years) or 4°C (2 years).
  • Molecular weight 341.13 g/mol and molecular formula C13H10BrFN2O3.
  • For research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-84212