Learn More
Sigma Aldrich Fine Chemicals Biosciences 2′,7′-Dichlorofluorescin diacetate, 4091-99-0, MFCD00128955, 50 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D688350MG
Encompass
Empirical Formula (Hill Notation): C24H16Cl2O7, Molecular Weight: 487.29, ≥97%, storage temp: −20°C, Synonym: 2′,7′-Dichlorodihydrofluorescein diacetate.
Catalog No. NC0844302