Learn More
Medchemexpress LLC 2-[(7-methoxy-4-methylquinazolin-2-yl)amino]-4,5-dimethyl-1h-pyrimidin-6-one | 374913-63-0 | MFCD02233199 | 99.2% | 311.34 g·mol⁻¹ | C16H17N5O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10023610MG
Encompass_Preferred
Madrasin is a small-molecule inhibitor of pre-mRNA splicing used for research. It prevents formation of splicing intermediates and products in vitro and interferes with early steps of spliceosome assembly; it can also modify splicing of endogenous pre-mRNA in cells. The compound is supplied as a crystalline powder and is typically characterized by high purity and a molecular weight of 311.34 g·mol⁻¹.
  • Potent pre-mRNA splicing inhibitor active in vitro.
  • Modulates endogenous pre-mRNA splicing in cells.
  • High purity suitable for biochemical and cellular assays.
  • Typically soluble in DMSO for stock preparation.
  • Supplied as a stable crystalline powder for storage and handling.
  • Intended for research use only, not for therapeutic use.
Catalog No. 50-002-15590