Learn More
Med Vet International 2.7 TPO/DPO Locking Plate 30 Right, J1212RB
Supplier: Med Vet International JOR1212RB
Triple Pelvic Osteotomy (TPO) and Double Pelvic Osteotomy (DPO) remain effrective techniques for the managment of hip dysplasia. Use of diverging, locking screws increases plate security as does the sightly larger footprint of our plates. Available in both 2.7 and 3.5 locking screw sizes at the following angles: 20.25 and 30 degrees. Extra long locking drill guides are available. 2.7mm TPO/DPO Locking Plate 30 Right.