Learn More
Medchemexpress LLC 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)prop-2-enoic acid | 261522-33-2 | MFCD09263343 | 99.6% | 309.32 | C18H15NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW348896100MG
Encompass_Preferred
Fmoc-Dha-OH is an Fmoc-protected dehydroalanine (Dha) amino-acid derivative supplied as a white to off-white solid. It is used as a building block in peptide chemistry and for installing dehydroalanine residues in peptides and proteins to enable conjugation, crosslinking, or mechanistic studies.
  • High purity suitable for research applications (99.6%).
  • White to off-white solid with defined storage recommendations.
  • Molecular weight 309.32, formula C18H15NO4.
  • Designed for use in solid-phase peptide synthesis workflows.
  • Enables installation of dehydroalanine residues for conjugation and mechanistic probes.
  • Stable when stored protected from light at refrigerated temperatures.
Catalog No. 50-002-82541