Learn More
Medchemexpress LLC 2-Acetamidomalonate (sodium) | 117976-12-2 | 98.0% | C₅H₅NNa₂O₅ | 250 MG

Supplier: Medchemexpress LLC 250MGHY79791
2-Acetamidomalonate (sodium) is a chemical compound identified by CAS No. 117976-12-2. It has a molecular formula of C₅H₅NNa₂O₅ and a molecular weight of 205.08. This product appears as a white to off-white solid with a purity of 98.0% by NMR and is intended for research use only.
- Appearance: White to off-white solid
- Purity (NMR): 98.0%
- Molecular formula: C₅H₅NNa₂O₅
- Molecular weight: 205.08
- Storage at 4°C: Sealed storage, away from moisture
- Storage in solvent at -80°C: 6 months
- Storage in solvent at -20°C: 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.