Learn More
Medchemexpress LLC 2-Acetamidomalonate sodium | 117976-12-2 | 98.0% | C5H5NNa2O5 | 10 G

Supplier: Medchemexpress LLC HY7979110G
2-Acetamidomalonate sodium is a chemical compound with the formula C5H5NNa2O5 and CAS number 117976-12-2. It is a drug intermediate for research use, typically appearing as a white to off-white solid.
- Purity of 98.0%
- Store at 4°C sealed, away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.