Learn More
Medchemexpress LLC 2-acetyl-N-[4-(hexafluoro-2-hydroxypropan-2-yl)phenyl]-5-methylsulfonyl-1,3-dihydroisoindole-1-carboxamide | 2101291-07-8 | 99.9% | 25 MG

Supplier: Medchemexpress LLC HY12038425MG
AZD-0284 is a selective inverse agonist of the nuclear receptor RORγ intended for laboratory research. It is supplied as a high-purity, white to off-white solid with defined physicochemical properties and recommended solubility and storage conditions.
- Selective inverse agonist of RORγ.
- Molecular formula C21H18F6N2O5S.
- Molecular weight 524.43 g/mol.
- Purity 99.9%.
- Appearance white to off-white solid.
- Soluble in DMSO (100 mg/mL); formulations provide ≥ 0.83 mg/mL in common vehicles.
- Storage: store cold; in solution, store under nitrogen at -80°C (6 months) or -20°C (1 month).
- For laboratory research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.