Learn More
Cayman Chemical 2-AcetylacteosIde 10mg
SDP
Supplier:  Cayman Chemical 3651110 MG
Encompass_Preferred
A phenylethanoid glycoside; selectively inhibits aldose reductase over maltase and sucrase (IC50s = 0.071, >300, and 277 µM, respectively, for the rat enzymes); scavenges superoxide radicals in a cell-free assay and inhibits AAPH-induced hemolysis in isolated rat red blood cells; decreases glutamate-induced cytotoxicity in primary rat cortical neurons at 0.1, 1, and 10 µM; protects primary mouse hepatocytes against D-galactosamine-induced cytotoxicity (IC50 = 4.8 UG/ml); prevents decreases in bone strength in ovariectomized mice from 10-40 mg/kg
Catalog No. 50-291-1620