Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose | 55637-63-3 | 98.0% | 383.35 g/mol | C14H25NO11 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW6990371MG

Encompass_Preferred

2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is a glycosylated N-acetylamino 2-deoxy sugar supplied as a white solid for research use in glycobiology, carbohydrate chemistry, and analytical applications. It is provided in small quantities for use as a reagent, reference standard, or synthetic intermediate.

  • White solid powder, suitable for laboratory storage and handling.
  • High purity: 98.0% by assay.
  • Molecular weight: 383.35 g/mol.
  • Chemical formula: C14H25NO11.
  • CAS number: 55637-63-3 for unambiguous identification.

Catalog No. 50-002-84160


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.