Learn More
Medchemexpress LLC 2-(acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose | 55637-63-3 | 98.0% | 383.35 g/mol | C14H25NO11 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW69903725MG
Encompass_Preferred
2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is an N-acetylated deoxy sugar derivative supplied as a solid biochemical reagent for glycobiology research. It is used to study carbohydrate structure, glycan biosynthesis, enzymology, and protein-glycan recognition.
  • High purity: 98.0%.
  • Molecular weight: 383.35 g/mol.
  • Solid powder formulation suitable for biochemical assays.
  • Powder stable at -20°C for up to 3 years; in solvent store at -80°C for up to 6 months.
  • Available in small quantities for research applications (e.g., 25 mg).
Catalog No. 50-002-84619