Learn More
Medchemexpress LLC HY-W015912 500mg Medchemexpress, 2-Acetylfuran CAS:1192-62-7 Purity:>98%
Supplier: Medchemexpress LLC HYW015912500MG
Medchemexpress, HY-W015912 500mg 2-Acetylfuran CAS:1192-62-7 2-Acetylfuran (2-Furyl methyl ketone), an important flavour compound or intermediate in foods, is isolated from essential oils, sweet corn products, fruits and flowers. 2-Acetylfuran also can be formed from glucose and glycine by Maillard reaction. 2-Acetylfuran can be used to synthesis Cefuroxime. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.